Merck
  • Recurring methicillin-resistant Staphylococcus aureus infections in a football team.
Recurring methicillin-resistant Staphylococcus aureus infections in a football team.
Emerging infectious diseases (2005-04-15)
Dao M Nguyen, Laurene Mascola, Elizabeth Brancoft
ABSTRACT
An outbreak of community-associated methicillin-resistant Staphylococcus aureus (MRSA) skin and soft tissue infection (SSTI) occurred in a college football team from August to September 2003. Eleven case-players were identified, and boils were the most common sign. Linemen had the highest attack rate (18%). Among 99 (93% of team) players with cultured specimens, 8 (8%) had positive MRSA nasal cultures. All available case-players' MRSA isolates characterized had the community-associated pulsed-field type USA300. A case-control study found that sharing bars of soap and having preexisting cuts or abrasions were associated with infection. A carrier-control study found that having a locker near a teammate with an SSTI, sharing towels, and living on campus were associated with nasal carriage. Successful outbreak control measures included daily hexachlorophene showers and hygiene education.