Merck
  • Reproductive effects in German cockroaches by ecdysteroid agonist RH-0345, juvenile hormone analogue methoprene and carbamate benfuracarb.
Reproductive effects in German cockroaches by ecdysteroid agonist RH-0345, juvenile hormone analogue methoprene and carbamate benfuracarb.
Communications in agricultural and applied biological sciences (2005-03-12)
A Maiza, S Kilani, J P Farine, G Smagghe, N Aribi, N Soltani
ABSTRACT
Blatta germanica is the more prevalent cockroach species in Algeria. In the present study, we tested the effect on reproduction in B. germanica of two insect growth regulators, RH-0345, a benzoylhydrazine analogue that mimics the action of 20-hydroxyecdysone, and methoprene, one of the most commercially important juvenile hormone analogues, and a novel carbamate insecticide, benfuracarb. The compounds were applied topically (10 and 20 microg/insect for RH-0345, and 1 and 10 microg/insect for methoprene) or orally administrated (at 2% for benfuracarb) on newly emerged females and evaluated on reproductive events during the adult life (2, 4 and 6 days). Treatment with RH-0345 and benfuracarb reduced significantly the number of oocytes, the size and the volume of the basal oocyte during the experimental period. Methoprene distorted the ovarian development since it caused a significant reduction in the number of oocytes at 2, 4 and 6 days for the two tested doses, and an increase in oocyte size at 2, 4 and 6 days with 1 microg and a decrease with 10 microg. In a second series of experiments, the effects of these compounds were assayed on the ovarian proteins. Data from biochemical analysis revealed that RH-0345 and benfuracarb reduced the ovarian amounts of proteins, while treatment with methoprene increased it during the sexual maturation.