Merck
  • Clindamycin is neuroprotective in experimental Streptococcus pneumoniae meningitis compared with ceftriaxone.
Clindamycin is neuroprotective in experimental Streptococcus pneumoniae meningitis compared with ceftriaxone.
Journal of neurochemistry (2004-12-09)
Tobias Böttcher, Hao Ren, Michel Goiny, Joachim Gerber, Jens Lykkesfeldt, Ulrich Kuhnt, Miriam Lotz, Stephanie Bunkowski, Carola Werner, Ingmar Schau, Annette Spreer, Stephan Christen, Roland Nau
ABSTRACT
In animal models of Streptococcus pneumoniae meningitis, rifampin is neuroprotective in comparison to ceftriaxone. So far it is not clear whether this can be generalized for other protein synthesis-inhibiting antimicrobial agents. We examined the effects of the bactericidal protein synthesis-inhibiting clindamycin (n = 12) on the release of proinflammatory bacterial components, the formation of neurotoxic compounds and neuronal injury compared with the standard therapy with ceftriaxone (n = 12) in a rabbit model of pneumococcal meningitis. Analysis of the CSF and histological evaluation were combined with microdialysis from the hippocampal formation and the neocortex. Compared with ceftriaxone, clindamycin reduced the release of lipoteichoic acids from the bacteria (p = 0.004) into the CSF and the CSF leucocyte count (p = 0.011). This led to lower extracellular concentrations of hydroxyl radicals (p = 0.034) and glutamate (p = 0.016) in the hippocampal formation and a subsequent reduction of extracellular glycerol levels (p = 0.018) and neuronal apoptosis in the dentate gyrus (p = 0.008). The present data document beneficial effects of clindamycin compared with ceftriaxone on various parameters linked with the pathophysiology of pneumococcal meningitis and development of neuronal injury. This study suggests neuroprotection to be a group effect of bactericidal protein synthesis-inhibiting antimicrobial agents compared with the standard therapy with beta-lactam antibiotics in meningitis.