Merck
  • Probing NMR parameters, structure and dynamics of 5-nitroimidazole derivatives. Density functional study of prototypical radiosensitizers.
Probing NMR parameters, structure and dynamics of 5-nitroimidazole derivatives. Density functional study of prototypical radiosensitizers.
Magnetic resonance in chemistry : MRC (2004-11-24)
Teodorico C Ramalho, Michael Bühl
ABSTRACT
The 15N chemical shifts of metronidazole (1), secnidazole (2), nimorazole (3) and tinidazole (4), radiosensitizers based on the 5-nitroimidazole motif, are reported. A detailed computational study of 1 is presented, calling special attention to the performance of various theoretical methods in reproducing the 13C and 15N data observed in solution. The most sophisticated approach involves density functional-based Car-Parrinello molecular dynamics simulations (CPMD) of 1 in aqueous solution (BP86 level) and averaging chemical shifts over snapshots from the trajectory. In the NMR calculations for these snapshots (performed at the B3LYP level), a small number of discrete water molecules are retained, and the remaining bulk solution effects are included via a polarizable continuum model (PCM). A similarly good accord with experiment is obtained from much less involved, static geometry optimization and NMR computation of pristine 1 employing a PCM approach. Solvent effects on delta(15N), which are of the order of up to 20 ppm, are not due to changes in geometric parameters upon solvation, but arise from the direct response of the electronic wavefunction to the presence of the solvent, which can be represented by discrete molecules and/or the dielectric bulk.