Merck
Stereochemical control of the Passerini reaction.
Organic letters (2004-11-05)
Peter R Andreana, Chang C Liu, Stuart L Schreiber
ABSTRACT
A catalytic asymmetric Passerini reaction using tridentate indan (pybox) Cu(II) Lewis acid complex 4 with substrates capable of bidentate coordination has been achieved. The reaction occurs via ligand-accelerated catalysis.