Merck
  • Oxyfluorfen toxic effect on S. obliquus evaluated by different photosynthetic and enzymatic biomarkers.
Oxyfluorfen toxic effect on S. obliquus evaluated by different photosynthetic and enzymatic biomarkers.
Archives of environmental contamination and toxicology (2004-01-08)
L Geoffroy, D Dewez, G Vernet, R Popovic
ABSTRACT
The effect of oxyfluorfen was investigated when alga Scenedesmus obliquus has been exposed to different concentrations (7.5, 15, and 22.5 microg x L(-1)) at 12, 24, and 48 hours of exposure. Toxicity test was done by using 13 biomarkers concerning growth rate, chlorophyll content and indicators of photosynthetic and antioxidant enzyme activities. The change of the 13 parameters showed a great variation of sensitivity indicating differences in parameters' suitability to be used as biomarkers when alga culture was exposed to oxyfluorfen toxicity. The order of sensitivity between those biomarkers was: Antenna size (ABS/RC) > Chlorophyll content > Catalase (CAT) > Operational PSII quantum yield (phiS(PSII)) > Glutathione S-transferase (GST) > Functional plastoquinone pool (Q(PQ)) > Glutathione reductase (GR) > Growth rate > Nonphotochemical quenching (QN) > Proton gradient quenching (Q(Emax)) > Ascorbate peroxidase (APX) > Photochemical quenching (Q(p)) > Maximum PSII quantum yield (Phi(PSII)). The effect of oxyfluorfen on the changes of those parameters was interpreted as a result of herbicide mode of action at molecular level of alga cellular system. This study indicated for some photosynthetic and enzymatic biomarkers to be useful indicators of toxicity effect induced in non-target alga species. Determination of biomarkers' sensitivity order may facilitate their selection to be used in environmental risk assessment of polluted water.