Merck
  • Organochalcogens effects on delta-aminolevulinate dehydratase activity from human erythrocytic cells in vitro.
Organochalcogens effects on delta-aminolevulinate dehydratase activity from human erythrocytic cells in vitro.
Toxicology (2003-09-11)
C W Nogueira, V C Borges, G Zeni, J B T Rocha
ABSTRACT
Organochalcogens are important intermediates and useful reagents in organic synthesis, which can increase human exposure risk to these chemicals in the workplace. As well, there are a number of reported cases of acute toxicity following organochalcogen ingestion of vitamins and dietary supplements. Since, the erythrocytic delta-ALA-D activity could be an important indicator of toxicity this report investigated the organochalcogens effects on blood delta-ALA-D in vitro. To investigate a possible involvement of cysteinyl groups in the inhibitory actions of diphenyl diselenide, diphenyl ditelluride and Ebselen (4-100 micro M), the effects of thiol reducing agents (0-3 mM) or zinc chloride (0-2 mM) were examined. Diphenyl ditelluride, diphenyl diselenide and Ebselen inhibited in a concentration-dependent manner delta-ALA-D activity from human erythrocytes. Ebselen was lesser delta-ALA-D inhibitor than (PhSe)(2) and (PhTe)(2), whereas the diorganoyldichalcogenides displayed similar inhibitory potency towards delta-ALA-D. Dithiothreitol, a hydrophobic SH-reducing agent, was able to reactivate and to protect inhibited delta-ALA-D. The pre-incubation of blood with the inhibitors changed considerably the reversing potency of thiols. From these findings we suggest that organochalcogens inactivate in vitro human erythrocyte delta-ALA-D by an interaction with the sulfhydryl group essential of the enzyme activity.