Merck
  • Catecholamines block the antimitogenic effect of estradiol on human glomerular mesangial cells.
Catecholamines block the antimitogenic effect of estradiol on human glomerular mesangial cells.
Hypertension (Dallas, Tex. : 1979) (2003-08-13)
Raghvendra K Dubey, Lefteris C Zacharia, Delbert G Gillespie, Bruno Imthurn, Edwin K Jackson
ABSTRACT
Local sequential conversion of estradiol to hydroxyestradiols and methoxyestradiols by CYP450 and catechol-O-methyltransferase, respectively, contributes to the antimitogenic effects of estradiol on glomerular mesangial cell growth via estrogen receptor-independent mechanisms. Catecholamines are also substrates for catechol-O-methyltransferase and therefore, might abrogate the renoprotective effects of estradiol by inhibiting formation of methoxyestradiols. To test this hypothesis, we investigated the antimitogenic effects of estradiol on human glomerular mesangial cell proliferation and collagen synthesis in the presence and absence of catecholamines. Norepinephrine, epinephrine, and isoproterenol abrogated the inhibitory effects of estradiol on cell number, DNA synthesis, and collagen synthesis. For example, serum-induced DNA synthesis was inhibited from 100% to 62+/-1.9% by 0.1 micromol/L estradiol, and these inhibitory effects were reversed to 91+/-1.9% by 1 micromol/L epinephrine, 90.7+/-3.3% by 1 micromol/L isoproterenol, 87.5+/-2.8% by 10 micromol/L norepinephrine, and 92+/-1% by 10 micromol/L OR486 (catechol-O-methyltransferase inhibitor). The interaction of catecholamines with estradiol was not affected by phentolamine or propanolol, alpha- and beta-adrenoceptor antagonists, respectively. Similar to estradiol, the antimitogenic effects of 2-hydroxyestradiol were abrogated by epinephrine, isoproterenol, and OR486. In contrast to estradiol and 2-hydroxyestradiol, the antimitogenic effects of 2-methoxyestradiol were not attenuated by epinephrine, isoproterenol, or OR486. Norepinephrine, epinephrine, and isoproterenol inhibited the conversion of both estradiol and 2-hydroxyestradiol to 2-methoxyestradiol. Our findings suggest that catecholamines within the glomeruli might abrogate the antimitogenic effects of estradiol by blocking the conversion of 2-hydroxyestradiol to 2-methoxyestradiol.