- Total synthesis of (+)-strictifolione.
Total synthesis of (+)-strictifolione.
Organic letters (2003-05-24)
Samir BouzBouz, Janine Cossy
PMID12762705
ABSTRACT
[reaction: see text] The synthesis of (+)-strictifolione was achieved from 3-phenylproprionaldehyde by using enantioselective allyltitanations to control the stereogenic centers at C6, C4', and C6' and a cross-methathesis to control the configuration of the double bond at C1'-C2'.