Merck
  • Opposite modulation of regulators of G protein signalling-2 RGS2 and RGS4 expression by dopamine receptors in the rat striatum.
Opposite modulation of regulators of G protein signalling-2 RGS2 and RGS4 expression by dopamine receptors in the rat striatum.
Neuroscience letters (2002-11-07)
Muriel Geurts, Emmanuel Hermans, Jean Marie Maloteaux
ABSTRACT
The role of dopaminergic transmission on striatal mRNA levels of regulators of G protein signalling proteins RGS 2-12 was evaluated by quantitative in situ hybridisation. A single injection of L-dopa (50 mg/kg i.p.) significantly increased the RGS2 mRNA level (by 25%), an effect that was specifically abolished by the D1 dopamine receptor antagonist R(+)-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride (R(+)-SCH23390), but not changed by S(-)-eticlopride. Interestingly, the administration of this D2 dopamine receptor antagonist alone markedly enhanced the expression of RGS2 (by 71%), which suggests a constitutive inhibition of RGS2 expression by D2 dopamine receptors. Opposite results were obtained concerning the regulation of RGS4 since L-dopa alone was without effect whereas co-administration of L-dopa and R(+)-SCH23390 significantly enhanced the RGS4 mRNA levels (by 38%). In conclusion, D1 and D2 dopamine receptors appear to mediate opposite regulatory effects on RGS2 and RGS4 expression in the striatum.