Merck
  • Use of potassium tellurite for testing the survival and viability of Pseudomonas pseudoalcaligenes KF707 in soil microcosms contaminated with polychlorinated biphenyls.
Use of potassium tellurite for testing the survival and viability of Pseudomonas pseudoalcaligenes KF707 in soil microcosms contaminated with polychlorinated biphenyls.
Research in microbiology (2002-09-18)
Giulio Zanaroli, Stefano Fedi, Monica Carnevali, Fabio Fava, Davide Zannoni
ABSTRACT
This study shows that the oxyanion tellurite TeO3(2-) can be used as a tool to detect and quantify the release in soil microcosms of Pseudomonas pseudoalcaligenes KF707, a strain spontaneously resistant to tellurite with a minimal inhibitory concentration (MIC) of 150 microg ml(-1). KF707 cells which carry the genes for degradation of a wide range of polychlorinated biphenyl congeners (PCBs) were used for inoculation of laboratory microcosms prepared with two different PCB-contaminated soils (Ci/s and Di/s) in the presence or absence of biphenyl as carbon source. In all microcosms supplemented with biphenyl, significant survival of strain KF707 was noted over a time period of 35 days; conversely, in microcosms containing Ci/s soil without biphenyl addition a rapid decrease in KF707 inoculated cells was observed. By comparing the number of inoculated KF707 cells with the number of indigenous bacteria growing on biphenyl (IBGB) of both Ci/s and Di/s microcosms, it could be concluded that the KF707/IBGB ratio is a relevant parameter in determining the fate of the added strain. The efficacy of potassium tellurite as a selective marker to monitor strain KF707 in laboratory microcosms was confirmed by ARDRA analyses of the 16S rDNA, while the isolated indigenous bacteria growing on biphenyl were identified as members of three different species of the genus Pseudomonas. We also report that in microcosms inoculated with KF707 cells in the absence of biphenyl, only low chlorinated biphenyls were degraded.
Product Description
Millipore
Potassium tellurite solution, 1% in H2O, suitable for microbiology