Merck
  • Effect of novel modulators of protein kinase C activity upon chemotherapy-induced differentiation and apoptosis in myeloid leukemic cells.
Effect of novel modulators of protein kinase C activity upon chemotherapy-induced differentiation and apoptosis in myeloid leukemic cells.
Anti-cancer drugs (2002-08-21)
Gerold Meinhardt, Elfriede Eppinger, Ralf Schmidmaier
ABSTRACT
Modulation of protein kinase C (PKC) activity has been demonstrated to either prevent or enhance drug-induced apoptosis in various tissue types. We tested four novel modulators of PKC activity in comparison to the phorbol ester 12-O-tetradecanoylphorbol-13-acetate (TPA) for the capability to affect differentiation, cell cycle progression and apoptosis in the human myeloid leukemia cell lines U937 and HL-60. Farnesyl thiotriazole (FTT) and N-(n-heptyl)-5-chloro-1-naphthalenesulfonamide (SC-10) are both direct activators of PKC, whereas 6-(2-(4-[(4-fluorophe-nyl)phenylmethylene]-1-piperidinyl)ethyl)-7-methyl-5H-thiazolo[3,2-a]pyrimidin-5-one (R59022) and [3-[2-[4-(bis-(4-fluorophenyl)methylene]piperidin-1-yl)ethyl]-2,3-dihydro-2-thioxo-4(1H)-quin-azolinone (R59949) are diacyl glycerol kinase inhibitors that activate PKC by enhancing the levels of the endogenous ligand diacyl glycerol. U937 cells displayed a slight reduction in the number of cells in G(2)/M cell cycle phase after exposure to FTT, SC-10, R59022 and R59949, respectively. In contrast, HL-60 cells demonstrated a largely unaltered cell cycle distribution. Whereas TPA treatment resulted in a strong induction of p21(WAF/CIP1), c-Fos and c-Jun levels, neither one of the novel PKC activators altered expression of these proteins. Consequently, we tested the ability of the activators to cause membrane translocation of PKC. While TPA treatment resulted in translocation of the PKC isoforms alpha, delta and epsilon, SC-10 and FTT failed to induce alterations in the PKC content of the membrane and cytosolic fractions, respectively. Expression of the beta(2)-integrin CD11c that is induced during TPA-mediated differentiation remained unaltered after exposure to SC-10 and was partly reduced after treatment with FTT. To further investigate the effect of these activators upon apoptosis in leukemic cells, HL-60 and U937 cells were treated with 1-beta-D-arabinofuranosylcytosine (Ara-C) or etoposide (VP-16). Whereas TPA strongly reduced apoptosis in Ara-C- or VP-16-treated U937 cells, little if any reduction was observed after pretreatment with either FTT, SC-10, R59022 or R59949, respectively, in these cells. In contrast, TPA enhanced apoptosis in Ara-C- or VP-16-treated HL-60 cells. Interestingly, FTT and SC-10 demonstrated a protective effect in Ara-C-treated HL-60 cells. Taken together, these data suggest that the novel PKC activators FTT, SC-10, R59022 and R59949 exhibit modest biological effects upon leukemic blast cells, and are not capable of enhancing the apoptotic response of these cells to cytotoxic drugs.