Merck
  • The metabolism of 4-trifluoromethoxyaniline and [13C]-4-trifluoromethoxyacetanilide in the rat: detection and identification of metabolites excreted in the urine by NMR and HPLC-NMR.
The metabolism of 4-trifluoromethoxyaniline and [13C]-4-trifluoromethoxyacetanilide in the rat: detection and identification of metabolites excreted in the urine by NMR and HPLC-NMR.
Journal of pharmaceutical and biomedical analysis (2002-06-01)
M Tugnait, E M Lenz, P Phillips, M Hofmann, M Spraul, J C Lindon, J K Nicholson, I D Wilson
ABSTRACT
A combination of 19F, 1H NMR and HPLC-NMR spectroscopic approaches have been used to quantify and identify the urinary-excreted metabolites of 4-trifluoromethoxyaniline (4-TFMeA) and its [13C]-labelled acetanilide following i.p. administration at 50 mg/kg to rats. The major metabolite excreted in the urine for both compounds was a sulphated ring-hydroxylated metabolite (either 2- or 3-trifluoromethyl-5-aminosulphate) which accounted for approximately 32.3% of the dose following the administration of 4-TFMeA and approximately 29.9% following dosing of the acetanilide. The trifluoromethoxy-substituent appeared to be metabolically stable, with no evidence of O-detrifluoromethylation. There was no evidence of the excretion of N-oxanilic acids in urine, of the type seen with 4-trifluoromethylaniline.