Merck
  • Mast cells involvement in the inflammation and fibrosis development of the TNBS-induced rat model of colitis.
Mast cells involvement in the inflammation and fibrosis development of the TNBS-induced rat model of colitis.
Scandinavian journal of gastroenterology (2002-03-28)
Xiang Xu, S Weksler-Zangen, A Pikarsky, O Pappo, D Wengrower, S C Bischoff, M Pines, A Rivkind, E Goldin, F Levi-Schaffer
ABSTRACT
Mast cells have been implicated in chronic inflammatory conditions resulting in fibrosis, such as Crohn disease. However, a link between inflammation, fibrosis and mast cells has not been demonstrated in human or animal intestinal diseases. This work was undertaken to analyze whether mast cells play a role in inflammation and fibrosis in the TNBS-induced rat colitis. Rats were rectally instilled 2,4,6,-trinitrobenzene sulfonic acid in ethanol, and immediately or 4 days later injected daily i.p. with nedocromil sodium, a mast cell stabilizer, compound 48/80, a mast cell activator, or saline. Rats were sacrificed 5 days post-TNBS, or on day 21. Intestinal inflammation and fibrosis were assessed by gross and histopathological evaluation. Colonic mast cell numbers (toluidine blue) and collagen (type I mRNA expression) were evaluated. Mast cell sonicate was added to rat colon fibroblasts. Fibroblast proliferation (3H-thymidine), collagen synthesis (3H-proline) and contractile activity (tridimensional collagen lattice contraction) were then assessed. Nedocromil reduced inflammation and fibrosis possibly by decreasing mast cell numbers and activation and consequent collagen production. Compound 48/80 slightly enhanced the severity of the disease by activating mast cells. Mast cells increased fibroblast proliferation, collagen production and contractile activity. Mast cells are involved in the gastrointestinal tract inflammation and fibrosis of the TNBS-colitis rats.