Merck
  • Pentopyranosyl oligonucleotide systems. Part 11: Systems with shortened backbones: (D)-beta-ribopyranosyl-(4'-->3')- and (L)-alpha-lyxopyranosyl-(4'-->3')-oligonucleotides.
Pentopyranosyl oligonucleotide systems. Part 11: Systems with shortened backbones: (D)-beta-ribopyranosyl-(4'-->3')- and (L)-alpha-lyxopyranosyl-(4'-->3')-oligonucleotides.
Bioorganic & medicinal chemistry (2001-09-13)
H Wippo, F Reck, R Kudick, M Ramaseshan, G Ceulemans, M Bolli, R Krishnamurthy, A Eschenmoser
ABSTRACT
The (L)-alpha-lyxopyranosyl-(4'-->3')-oligonucleotide system-a member of a pentopyranosyl oligonucleotide family containing a shortened backbone-is capable of cooperative base-pairing and of cross-pairing with DNA and RNA. In contrast, corresponding (D)-beta-ribopyransoyl-(4'-->3')-oligonucleotides do not show base-pairing under similar conditions. We conclude that oligonucleotide systems can violate the 'six-bonds-per-backbone-unit' rule by having five bonds instead, if their vicinally bound phosphodiester bridges can assume an antiperiplanar conformation. An additional structural feature that seems relevant to the cross-pairing capability of the (L)-alpha-lyxopyranosyl-(4'-->3')-oligonucleotide system is its (small) backbone/basepair axes inclination. An inclination which is similar to that in B-DNA seems to be a prerequisite for an oligonucleotide system's capability to cross-pair with DNA.