- Cardenolide and oxypregnane glycosides from the root of Asclepias incarnata L.
Cardenolide and oxypregnane glycosides from the root of Asclepias incarnata L.
Chemical & pharmaceutical bulletin (2000-04-27)
T Warashina, T Noro
PMID10783071
ABSTRACT
Twenty-nine new oxypregnane glycosides were obtained along with two known cardenolides, frugoside and gofruside, and three known 12-O-acylated pregnane glycosides from the roots of Asclepias incarnata L. (Asclepiadaceae). By detailed studies of the 1H- and 13C-NMR spectra, the structures were determined to be tri- to penta glycosides of isolineolon, 12-O-acetyllineolon, ikemagenin, 12-O-benzoylisolineolon, and two new 12-O-acylated pregnanes.