Merck
  • Thiolated recombinant human tumor necrosis factor-alpha protects against Plasmodium berghei K173-induced experimental cerebral malaria in mice.
Thiolated recombinant human tumor necrosis factor-alpha protects against Plasmodium berghei K173-induced experimental cerebral malaria in mice.
Antimicrobial agents and chemotherapy (1999-05-01)
N S Postma, R C Hermsen, D J Crommelin, W M Eling, J Zuidema
ABSTRACT
The introduction of reactive thiol groups in recombinant human tumor necrosis factor (TNF) alpha (rhTNF-alpha) by the reagent succinimidyl-S-acetylthioacetate resulted in the formation of a chemically stabilized rhTNF-alpha trimer (rhTNFalpha-AT; as determined by sodium dodecyl sulfate-polyacrylamide gel electrophoresis analysis). rhTNFalpha-AT showed a substantially enhanced protective efficacy against the development of experimental murine cerebral malaria (ECM) after intravenous injection compared to the protective efficacy of nonmodified rhTNF-alpha. Administration of thiolated rhTNF-alpha with protected thiol groups (rhTNFalpha-ATA; no stabilized trimers in vitro) exhibited the same protective efficacy against ECM, while in vitro bioactivity was reduced. Parasitemia was significantly suppressed in rhTNF-treated mice that were protected against ECM but not in treated mice that developed ECM. Protection against ECM was not related to increased concentrations in plasma of soluble TNF receptor 1 and 2 directly after injection or at the moment of development of ECM in nontreated mice. The results indicate that thiolation of rhTNF-alpha leads to the formation of stable trimers with increased potential in vivo.