Merck
  • Development of a fluorescent indicator for nitric oxide based on the fluorescein chromophore.
Development of a fluorescent indicator for nitric oxide based on the fluorescein chromophore.
Chemical & pharmaceutical bulletin (1998-03-21)
H Kojima, K Sakurai, K Kikuchi, S Kawahara, Y Kirino, H Nagoshi, Y Hirata, T Nagano
ABSTRACT
Endogenous nitric oxide (NO) appears to modulate many physiological and pathophysiological processes. In order to obtain direct evidence for NO functions in vivo, we have developed 4,5-diaminofluorescein (DAF-2) as a novel fluorescent indicator for NO. Green-fluorescent triazolofluorescein formed by the reaction of NO and DAF-2 affords high sensitivity for NO (detection limit: 5 nM). Membrane-permeable DAF-2 diacetate (DAF-2 DA) was loaded into activated rat aortic smooth muscle cells, where the ester bonds are hydrolyzed by intracellular esterase, generating DAF-2. The fluorescence in the cells increased in a NO concentration-dependent manner. This imaging method should be useful for studies of the dynamic biological actions of NO at the molecular level with fine temporal and spatial resolution.