- Primary structure of alpha-clostripain light chain.
Primary structure of alpha-clostripain light chain.
European journal of biochemistry (1984-12-17)
A M Gilles, A Lecroisey, B Keil
PMID6391922
ABSTRACT
The primary structure of light chain of alpha-clostripain was determined by sequence analysis of peptides derived from tryptic digests purified by reverse-phase high-performance liquid chromatography. The 22 isolated tryptic peptides were aligned by peptides derived from chymotryptic and staphylococcal V8 proteinase digests. The light chain contains 133 amino acids residues and has a relative molecular mass of 15400. The prediction of its secondary structure is given.