Merck
  • Identification of a high-affinity orphanin FQ/nociceptin(1-11) binding site in mouse brain.
Identification of a high-affinity orphanin FQ/nociceptin(1-11) binding site in mouse brain.
Synapse (New York, N.Y.) (1999-10-19)
J P Mathis, I E Goldberg, S R Letchworth, J P Ryan-Moro, G W Pasternak
ABSTRACT
The presence of pairs of basic amino acids within the orphanin FQ/Nociceptin (OFQ/N) sequence has raised the possibility that truncated versions of the peptide might be physiologically important. OFQ/N(1-11) is pharmacologically active in mice, despite its poor affinity in binding assays (K(i) > 250 nM) for the OFQ/N receptor. Using an analog of OFQ/N(1-11), [(125)I][Tyr(10)]OFQ/N(1-11), we identified a high-affinity binding site (K(D) 234 pM; B(max) 43 fmol/mg protein) with a selectivity profile distinct from the OFQ/N receptor and all the traditional opioid receptors. This site had very high affinity for OFQ/N and its related peptides. The most striking differences between the new site and the OFQ/N receptor previously observed in brain were seen with traditional opioids. Dynorphin A analogs and alpha-neoendorphin competed with [(125)I][Tyr(10)]OFQ/N(1-11) binding in mouse brain with K(i) values below 10 nM, while naloxone benzoylhydrazone (K(i) 3.9 nM) labeled the [(125)I][Tyr(10)]OFQ/N(1-11) binding site as potently as many traditional opioid receptors. Several other opioids, including fentanyl, (-)cyclazocine, levallorphan, naltrindole, and diprenorphine, also displayed moderate affinities for this site. Finally, the [(125)I][Tyr(10)]OFQ/N(1-11) site had a unique regional distribution consistent with a distinct receptor. Thus, [(125)I][Tyr(10)]OFQ/N(1-11) labels a novel site in brain with a selectivity profile intermediate between that of either opioid or OFQ/N receptors.