Merck
  • Binding characteristics of naftopidil and alpha 1-adrenoceptor antagonists to prostatic alpha-adrenoceptors in benign prostatic hypertrophy.
Binding characteristics of naftopidil and alpha 1-adrenoceptor antagonists to prostatic alpha-adrenoceptors in benign prostatic hypertrophy.
Life sciences (1992-01-01)
S Yamada, M Suzuki, Y Kato, R Kimura, R Mori, K Matsumoto, M Maruyama, K Kawabe
ABSTRACT
Binding properties of naftopidil and alpha 1-adrenoceptor antagonists to alpha-adrenoceptors in prostates from benign prostatic hypertrophy (BPH) were characterized by radioreceptor assays using [3H]prazosin and [3H]rauwolscine. Specific binding of [3H]prazosin and [3H]rauwolscine in human prostatic membranes was saturable and of high affinity, and it showed a pharmacological specificity which characterized alpha 1 and alpha 2-adrenoceptors, respectively. Naftopidil and several alpha 1 antagonists competed for prostatic [3H]prazosin binding in order: R-(-)-YM-12617 greater than prazosin greater than bunazosin greater than terazosin greater than naftopidil greater than urapidil, and the inhibitory effect (Ki = 11.6 nM) of naftopidil was 10 to 45 times less potent than quinazoline derivatives such as prazosin, bunazosin and terazosin. The potencies of these antagonists in competing for [3H]prazosin binding sites in human prostates correlated well with their pharmacological potencies (pA2). Scatchard analysis indicated that the decrease of prostatic [3H]prazosin binding by naftopidil was due to a marked increase in the Kd value without a change in the Bmax value. The inhibition of prostatic [3H]prazosin binding by naftopidil was reversible. Naftopidil also inhibited prostatic [3H]rauwolscine binding (Ki = 70.0 nM). Thus, it is suggested that naftopidil antagonizes alpha 1-adrenoceptors in human prostates in a competitive and reversible manner.