Merck
  • Biological profile of L-745,870, a selective antagonist with high affinity for the dopamine D4 receptor.
Biological profile of L-745,870, a selective antagonist with high affinity for the dopamine D4 receptor.
The Journal of pharmacology and experimental therapeutics (1997-11-14)
S Patel, S Freedman, K L Chapman, F Emms, A E Fletcher, M Knowles, R Marwood, G Mcallister, J Myers, N Curtis, J J Kulagowski, P D Leeson, M Ridgill, M Graham, S Matheson, D Rathbone, A P Watt, L J Bristow, N M Rupniak, E Baskin, J J Lynch, C I Ragan
ABSTRACT
L-745,870,(3-([4-(4-chlorophenyl)piperazin-1-yl]methyl)-1H- pyrollo[2,3-b] pyridine, was identified as a selective dopamine D4 receptor antagonist with excellent oral bioavailability and brain penetration. L-745,870 displaced specific binding of 0.2 nM [3H] spiperone to cloned human dopamine D4 receptors with a binding affinity (Ki) of 0. 43 nM which was 5- and 20-fold higher than that of the standard antipsychotics haloperidol and clozapine, respectively. L-745,870 exhibited high selectivity for the dopamine D4 receptor (>2000 fold) compared to other dopamine receptor subtypes and had moderate affinity for 5HT2, sigma and alpha adrenergic receptors(IC50 < 300 nM). In vitro, L-745,870 (0.1-1 microM) exhibited D4 receptor antagonist activity, reversing dopamine (1 microM) mediated 1) inhibition of adenylate cyclase in hD4HEK and hD4CHO cells; 2) stimulation of [35S] GTPgammaS binding and 3) stimulation of extracellular acidification rate, but did not exhibit any significant intrinsic activity in these assays. Although standard antipsychotics increase dopamine metabolism or plasma prolactin levels in rodents, L-745,870 (</=30 mg/kg p.o.) had no effect in these assays. The lack of a suitable in vivo assay for D4 receptor activation prompted the use of in vivo surrogate marker assays which confirmed that doses of 5-60 microg/kg L-745,870 would be sufficient to occupy 50% D4 receptors in the brain. These results show that dopamine D4 receptor antagonism in the brain does not result in the same neurochemical consequences (increased dopamine metabolism or hyperprolactinemia) observed with typical neuroleptics.