Merck
  • Neuroleptics of the diphenylbutylpiperidine series are potent calcium channel inhibitors.
Neuroleptics of the diphenylbutylpiperidine series are potent calcium channel inhibitors.
Proceedings of the National Academy of Sciences of the United States of America (1986-10-01)
J P Galizzi, M Fosset, G Romey, P Laduron, M Lazdunski
ABSTRACT
[3H]Fluspirilene, a neuroleptic molecule of the diphenylbutylpiperidine series, binds to skeletal muscle transverse tubule membranes with a high affinity corresponding to a Kd of 0.11 +/- 0.04 nM, A 1:1 stoichiometry was found between [3H]fluspirilene binding and the binding of (-)-[3H]desmethoxyverapamil [(-)[3H]D888], one of the most potent Ca2+ channel inhibitors. Ca2+ channel inhibitors such as D888, verapamil, gallopamil, bepridil, or diltiazem antagonize [3H]fluspirilene binding besides antagonizing (-)[3H]-D888 binding. Neuroleptics, especially those of the diphenylbutylpiperidine family, also antagonize both (-)[3H]D888 binding and [3H]fluspirilene binding. There is an excellent correlation between affinities found from [3H]fluspirilene binding experiments and those found from (-)[3H]D888 binding experiments. Analysis of the properties of these cross-inhibitions indicates that [3H]fluspirilene binds to a site that is not identical to that for phenylalkylamine derivatives (gallopamil, verapamil, diltiazem, and bepridil). Voltage-clamp experiments have shown that fluspirilene is an efficient inhibitor of the voltage dependent Ca2+ channel, achieving a half-maximal effect near 0.1-0.2 nM and nearly complete blockade at 1 nM. Fluspirilene blockade has little voltage dependence.