- Complexes of the copper-containing amine oxidase from Arthrobacter globiformis with the inhibitors benzylhydrazine and tranylcypromine.
Complexes of the copper-containing amine oxidase from Arthrobacter globiformis with the inhibitors benzylhydrazine and tranylcypromine.
Acta crystallographica. Section F, Structural biology and crystallization communications (2008-07-09)
David B Langley, Daniel M Trambaiolo, Anthony P Duff, David M Dooley, Hans C Freeman, J Mitchell Guss
PMID18607080
ABSTRACT
Complexes of Arthrobacter globiformis amine oxidase (AGAO) with the inhibitors benzylhydrazine and tranylcypromine (an antidepressant drug) have been refined at 1.86 and 1.65 A resolution, respectively. Both inhibitors form covalent adducts with the TPQ cofactor. A tyrosine residue, proposed to act as a gate to the AGAO active site, is in its open conformation.