Merck
  • Cellular arachidonate-releasing function of novel classes of secretory phospholipase A2s (groups III and XII).
Cellular arachidonate-releasing function of novel classes of secretory phospholipase A2s (groups III and XII).
The Journal of biological chemistry (2003-01-11)
Makoto Murakami, Seiko Masuda, Satoko Shimbara, Sofiane Bezzine, Michael Lazdunski, Gérald Lambeau, Michael H Gelb, Satoshi Matsukura, Fumio Kokubu, Mitsuru Adachi, Ichiro Kudo
ABSTRACT
Here we report cellular arachidonate (AA) release and prostaglandin (PG) production by novel classes of secretory phospholipase A(2)s (sPLA(2)s), groups III and XII. Human group III sPLA(2) promoted spontaneous AA release, which was augmented by interleukin-1, in HEK293 transfectants. The central sPLA(2) domain alone was sufficient for its in vitro enzymatic activity and for cellular AA release at the plasma membrane, whereas either the unique N- or C-terminal domain was required for heparanoid-dependent action on cells to augment AA release, cyclooxygenase-2 induction, and PG production. Group III sPLA(2) was constitutively expressed in two human cell lines, in which other sPLA(2)s exhibited different stimulus inducibility. Human group XII sPLA(2) had a weak enzymatic activity in vitro and minimally affects cellular AA release and PG production. Cells transfected with group XII sPLA(2) exhibited abnormal morphology, suggesting a unique functional aspect of this enzyme. Based on the present results as well as our current analyses on the group I/II/V/X sPLA(2)s, general properties of cellular actions of a full set of mammalian sPLA(2)s in regulating AA metabolism are discussed.