Merck
  • Succinylcholine-induced hyperkalemia in acquired pathologic states: etiologic factors and molecular mechanisms.
Succinylcholine-induced hyperkalemia in acquired pathologic states: etiologic factors and molecular mechanisms.
Anesthesiology (2006-01-06)
J A Jeevendra Martyn, Martina Richtsfeld
ABSTRACT
Lethal hyperkalemic response to succinylcholine continues to be reported, but the molecular mechanisms for the hyperkalemia have not been completely elucidated. In the normal innervated mature muscle, the acetylcholine receptors (AChRs) are located only in the junctional area. In certain pathologic states, including upper or lower motor denervation, chemical denervation by muscle relaxants, drugs, or toxins, immobilization, infection, direct muscle trauma, muscle tumor, or muscle inflammation, and/or burn injury, there is up-regulation (increase) of AChRs spreading throughout the muscle membrane, with the additional expression of two new isoforms of AChRs. The depolarization of these AChRs that are spread throughout the muscle membrane by succinylcholine and its metabolites leads to potassium efflux from the muscle, leading to hyperkalemia. The nicotinic (neuronal) alpha7 acetylcholine receptors, recently described to be expressed in muscle also, can be depolarized not only by acetylcholine and succinylcholine but also by choline, persistently, and possibly play a critical role in the hyperkalemic response to succinylcholine in patients with up-regulated AChRs.