- Medium for use in antibiotic susceptibility testing of anaerobic bacteria.
Medium for use in antibiotic susceptibility testing of anaerobic bacteria.
Antimicrobial agents and chemotherapy (1976-12-01)
T D Wilkins, S Chalgren
PMID1008552
ABSTRACT
A medium is described which was designed for use in testing the minimal inhibitory concentration of antibiotics for anaerobic bacteria by agar dilution. It contains: Trypticase (1%), Gelysate (1%), yeast extract (0.5%), glucose (0.1%), pyruvate (0.1%), arginine (0.1%), NaCl (0.5%), hemin (5 mug/ml), vitamin K(1) (0.5 mug/ml), agar (1.5%). The medium does not require the addition of blood to support growth of most clinical isolates of anaerobic bacteria.