Merck
  • Role of advanced glycation endproducts and glyoxalase I in diabetic peripheral sensory neuropathy.
Role of advanced glycation endproducts and glyoxalase I in diabetic peripheral sensory neuropathy.
Translational research : the journal of laboratory and clinical medicine (2012-04-17)
Megan Jack, Douglas Wright
ABSTRACT
Diabetic neuropathy is the most common and debilitating complication of diabetes mellitus with more than half of all patients developing altered sensation as a result of damage to peripheral sensory neurons. Hyperglycemia results in altered nerve conduction velocities, loss of epidermal innervation, and development of painful or painless signs and symptoms in the feet and hands. Current research has been unable to determine whether a patient will develop insensate or painful neuropathy or be protected from peripheral nerve damage all together. One mechanism that has been recognized to have a role in the pathogenesis of sensory neuron damage is the process of reactive dicarbonyls forming advanced glycation endproducts (AGEs) as a direct result of hyperglycemia. The glyoxalase system, composed of the enzymes glyoxalase I (GLO1) and glyoxalase II, is the main detoxification pathway involved in breaking down toxic reactive dicarbonyls before producing carbonyl stress and forming AGEs on proteins, lipids, or nucleic acids. This review discusses AGEs, GLO1, their role in diabetic neuropathy, and potential therapeutic targets of the AGE pathway.