Merck
  • Immunohistological analysis in diagnosis of plasma cell myeloma based on cytoplasmic kappa/lambda ratio of CD38-positive plasma cells.
Immunohistological analysis in diagnosis of plasma cell myeloma based on cytoplasmic kappa/lambda ratio of CD38-positive plasma cells.
Hematology (Amsterdam, Netherlands) (2012-11-22)
Shoko Nakayama, Taiji Yokote, Yuji Hirata, Kazuki Iwaki, Toshikazu Akioka, Takuji Miyoshi, Uta Nishiwaki, Yuki Masuda, Nobuya Hiraoka, Ayami Takayama, Yasuichiro Nishimura, Motomu Tsuji, Toshiaki Hanafusa
ABSTRACT
The accurate determination of cytoplasmic immunoglobulin (cIg) light chain (LC) expression is important to differentiate reactive plasmacytosis from a clonal plasma cell neoplasm such as plasma cell myeloma (PCM). Through retrospective analysis, we studied the cytoplasmic kappa/lambda ratio of CD38-positive plasma cells in the bone marrow from 19 PCM patients and 19 controls. To demonstrate cIg LC expression, the bone marrow was immunostained for IgA, IgG, IgM, kappa, and lambda. The kappa/lambda ratio was defined as the ratio of the kappa-positive cell to the lambda-positive cell in plasma cells. PCM cells were distinguished from normal plasma cells by cut-off levels between 0.59 and 4.0, a sensitivity of 94.7%, and a specificity of 94.7%. The detection of the cytoplasmic kappa/lambda ratio of CD38-positive plasma cells may be a useful tool in the diagnosis of PCM and the correct diagnosis of PCM may be achieved more simply.