Merck
  • Cathepsin D deficiency induces cytoskeletal changes and affects cell migration pathways in the brain.
Cathepsin D deficiency induces cytoskeletal changes and affects cell migration pathways in the brain.
Neurobiology of disease (2012-10-17)
Sabine Koch, Enzo Scifo, Anne Rokka, Pascal Trippner, Maria Lindfors, Reeta Korhonen, Garry L Corthals, Ismo Virtanen, Maciej Lalowski, Jaana Tyynelä
ABSTRACT
Cathepsin D deficiency is a fatal neurodegenerative disease characterized by extreme loss of neurons and myelin. Our previous studies have demonstrated that structural and functional alterations in synapses are central to the disease pathogenesis. Therefore, we took a systematic approach to examine the synaptic proteome in cathepsin D knock-out mice, where the synaptic pathology resembles that of human patients. We applied quantitative mass spectrometry analysis on synaptosomal fractions isolated from cathepsin D knock-out and control mice at the age of 24 days. From the approximately 600 identified proteins, 43 were present in different amounts (P<0.05, measured in triple biological replicates) in cathepsin D knock-out mice compared to controls. We connected and bridged these 43 proteins using protein interaction data, and overlaid the network with brain specific gene expression information. Subsequently, we superimposed the network with Gene Ontology, pathway, phenotype and disease involvement, allowing construction of a dynamic, disease-protein centered network and prediction of functional modules. The measured changes in the protein levels, as well as some of the bioinformatically predicted ones, were confirmed by quantitative Western blotting or qualitative immunohistochemistry. This combined approach indicated alterations in distinct cellular entities, previously not associated with the disease, and including microtubule associated cytoskeleton and cell projection organization. Cell spreading and wound healing assays confirmed strongly compromised spatial orientation, associated with changes in distribution of focal adhesions and integrin assembly, in cathepsin D deficient cells. These changes might contribute to commencement of synaptic alterations and neuronal degeneration observed in cathepsin D deficiency.