Merck
  • Protein solubility and differential proteomic profiling of recombinant Escherichia coli overexpressing double-tagged fusion proteins.
Protein solubility and differential proteomic profiling of recombinant Escherichia coli overexpressing double-tagged fusion proteins.
Microbial cell factories (2010-08-31)
Chung-Hsien Cheng, Wen-Chien Lee
ABSTRACT
Overexpression of recombinant proteins usually triggers the induction of heat shock proteins that regulate aggregation and solubility of the overexpressed protein. The two-dimensional gel electrophoresis (2-DE)-mass spectrometry approach was used to profile the proteome of Escherichia coli overexpressing N-acetyl-D-glucosamine 2-epimerase (GlcNAc 2-epimerase) and N-acetyl-D-neuraminic acid aldolase (Neu5Ac aldolase), both fused to glutathione S-transferase (GST) and polyionic peptide (5D or 5R). Overexpression of fusion proteins by IPTG induction caused significant differential expression of numerous cellular proteins; most of these proteins were down-regulated, including enzymes connected to the pentose phosphate pathway and the enzyme LuxS that could lead to an inhibition of tRNA synthesis. Interestingly, when plasmid-harboring cells were cultured in LB medium, gluconeogenesis occurred mainly through MaeB, while in the host strain, gluconeogenesis occurred by a different pathway (by Mdh and PckA). Significant up-regulation of the chaperones ClpB, HslU and GroEL and high-level expression of two protective small heat shock proteins (IbpA and IbpB) were found in cells overexpressing GST-GlcNAc 2-epimerase-5D but not in GST-Neu5Ac aldolase-5R-expressing E. coli. Although most of the recombinant protein was present in insoluble aggregates, the soluble fraction of GST-GlcNAc 2-epimerase-5D was higher than that of GST-Neu5Ac aldolase-5R. Also, in cells overexpressing recombinant GST-GlcNAc 2-epimerase-5D, the expression of σ32 was maintained at a higher level following induction. Differential expression of metabolically functional proteins, especially those in the gluconeogenesis pathway, was found between host and recombinant cells. Also, the expression patterns of chaperones/heat shock proteins differed among the plasmid-harboring bacteria in response to overproduction of recombinant proteins. In conclusion, the solubility of overexpressed recombinant proteins could be enhanced by maintaining the expression of σ32, a bacterial heat shock transcription factor, at higher levels during overproduction.