Merck
  • Cloning of dextransucrase gene from Leuconostoc citreum HJ-P4 and its high-level expression in E. coli by low temperature induction.
Cloning of dextransucrase gene from Leuconostoc citreum HJ-P4 and its high-level expression in E. coli by low temperature induction.
Journal of microbiology and biotechnology (2009-09-08)
Ah-Rum Yi, So-Ra Lee, Myoung-Uoon Jang, Jung-Mi Park, Hyun-Ju Eom, Nam Soo Han, Tae-Jip Kim
ABSTRACT
A dextransucrase (LcDS) gene from Leuconostoc citreum HJ-P4 has been amplified and cloned in E. coli. The LcDS gene consists of 4,431 nucleotides encoding 1,477 amino acid residues sharing 63-98% of amino acid sequence identities with other known dextransucrases from Leuc. mesenteroides. Interestingly, 0.1 mM of IPTG induction at 15 degrees remarkably increased the LcDS productivity to 19,187 U/l culture broth, which was over 330-fold higher than that induced at 37 degrees. Optimal reaction temperature and pH of LcDS were determined as 35 degrees and pH 5.5 in 20 mM sodium acetate buffer, respectively. Meanwhile, 0.1 mM CaCl(2) increased its activity to the maximum of 686 U/mg, which was 2.1-fold higher than that in the absence of calcium ion. Similar to the native Leuconostoc dextransucrase, recombinant LcDS could successfully produce a series of isomaltooligosaccharides from sucrose and maltose, on the basis of its transglycosylation activity.