Merck
  • Trypanosoma cruzi: altered parasites after in vitro treatment with gangliosides, a therapeutic agent in experimental Chagas' disease.
Trypanosoma cruzi: altered parasites after in vitro treatment with gangliosides, a therapeutic agent in experimental Chagas' disease.
Experimental parasitology (2009-04-09)
S Cossy Isasi, M Rodríguez, B M I Pereira, C Díaz-Luján, R E Fretes, D I Haüen
ABSTRACT
Biochemical and structural modifications were investigated in axenic cultured Trypanosoma cruzi after treatment with gangliosides. Fluorescence anisotropy showed dose dependent increments in parasite membranes of ganglioside treated epimastigotes. NADP-GDH activity increased in parasites treated at day 4 (13%), 7 (137.2%), and 14 (28.50%) while NAD-MDH but decreased from day 7 to 21 (-5.74%, -32.22%, -27.92%). Treated parasites presented electron-lucent vacuoles opposite to the cytostoma, multilamellar bodies and dilated mitochondrion cristae, disorganized kinetoplast and altered heterochromatin structure. Gangliosides inhibited fusogenic ability (80%) and PLA2 activity (>75%) from the parasite. The same occurred with anti-PLA2 antibodies. Trypomastigotes suffered loss of cytoplasmic material and organelles when GM1 was present in culture medium. We propose that exogenous gangliosides produced: altered lipid order, inhibited membrane enzymes, the parasite energy source shifted from glucose to amino acids, ending on a structural transformation which signals parasite cell death.