Merck
  • Cloning and expression of the sucrose phosphorylase gene from Leuconostoc mesenteroides in Escherichia coli.
Cloning and expression of the sucrose phosphorylase gene from Leuconostoc mesenteroides in Escherichia coli.
Biotechnology letters (2007-11-27)
Jin-Ha Lee, Young-Hwan Moon, Nahyun Kim, Young-Min Kim, Hee-Kyoung Kang, Ji-Yeon Jung, Emad Abada, Seong-Soo Kang, Doman Kim
ABSTRACT
The gene encoding sucrose phosphorylase (742sp) in Leuconostoc mesenteroides NRRL B-742 was cloned and expressed in Escherichia coli. The nucleotide sequence of the transformed 742sp comprised an ORF of 1,458 bp giving a protein with calculated molecular mass of 55.3 kDa. 742SPase contains a C-terminal amino acid sequence that is significantly different from those of other Leu. mesenteroides SPases. The purified 742SPase had a specific activity of 1.8 U/mg with a K (m) of 3 mM with sucrose as a substrate; optimum activity was at 37 degrees C and pH 6.7. The purified 742SPase transferred the glucosyl moiety of sucrose to cytosine monophosphate (CMP).