Merck
  • The role of cyclic nucleotide phosphodiesterases in the regulation of adipocyte lipolysis.
The role of cyclic nucleotide phosphodiesterases in the regulation of adipocyte lipolysis.
Journal of lipid research (2004-12-18)
Peter B Snyder, James M Esselstyn, Kate Loughney, Sharon L Wolda, Vincent A Florio
ABSTRACT
This study assessed the effects of selective inhibitors of 3',5'-cyclic nucleotide phosphodiesterases (PDEs) on adipocyte lipolysis. IC224, a selective inhibitor of type 1 phosphodiesterase (PDE1), suppressed lipolysis in murine 3T3-L1 adipocytes (69.6 +/- 5.4% of vehicle control) but had no effect in human adipocytes. IC933, a selective inhibitor of PDE2, had no effect on lipolysis in either cultured murine 3T3-L1 adipocytes or human adipocytes. Inhibition of PDE3 with cilostamide moderately stimulated lipolysis in murine 3T3-L1 and rat adipocytes (397 +/- 25% and 235 +/- 26% of control, respectively) and markedly stimulated lipolysis in human adipocytes (932 +/- 7.6% of control). Inhibition of PDE4 with rolipram moderately stimulated lipolysis in murine 3T3-L1 adipocytes (291 +/- 13% of control) and weakly stimulated lipolysis in rat adipocytes (149 +/- 7.0% of control) but had no effect on lipolysis in human adipocytes. Cultured adipocytes also responded differently to a combination of PDE3 and PDE4 inhibitors. Simultaneous exposure to cilostamide and rolipram had a synergistic effect on lipolysis in murine 3T3-L1 and rat adipocytes but not in human adipocytes. Hence, the relative importance of PDE3 and PDE4 in regulating lipolysis differed in cultured murine, rat, and human adipocytes.