Merck
  • Enzymatic preparation of metabolic intermediates, 3-dehydroquinate and 3-dehydroshikimate, in the shikimate pathway.
Enzymatic preparation of metabolic intermediates, 3-dehydroquinate and 3-dehydroshikimate, in the shikimate pathway.
Bioscience, biotechnology, and biochemistry (2006-12-08)
Osao Adachi, Yoshitaka Ano, Hirohide Toyama, Kazunobu Matsushita
ABSTRACT
A method for enzymatic preparation of 3-dehydroquinate and 3-dehydroshikimate in the shikimate pathway was established by controlling the enzyme activity of 3-dehydroquinate dehydratase. When quinate was incubated with the membrane fraction of acetic acid bacteria at pH 5.0, 3-dehydroquinate was formed as the predominant product. 3-Dehydroshikimate was the sole product when incubated at pH 8.0. Mutual separation of the metabolic intermediates was also exemplified.