Merck
  • TSPYL5 SNPs: association with plasma estradiol concentrations and aromatase expression.
TSPYL5 SNPs: association with plasma estradiol concentrations and aromatase expression.
Molecular endocrinology (Baltimore, Md.) (2013-03-23)
Mohan Liu, James N Ingle, Brooke L Fridley, Aman U Buzdar, Mark E Robson, Michiaki Kubo, Liewei Wang, Anthony Batzler, Gregory D Jenkins, Tracy L Pietrzak, Erin E Carlson, Matthew P Goetz, Donald W Northfelt, Edith A Perez, Clark V Williard, Daniel J Schaid, Yusuke Nakamura, Richard M Weinshilboum
ABSTRACT
We performed a discovery genome-wide association study to identify genetic factors associated with variation in plasma estradiol (E2) concentrations using DNA from 772 postmenopausal women with estrogen receptor (ER)-positive breast cancer prior to the initiation of aromatase inhibitor therapy. Association analyses showed that the single nucleotide polymorphisms (SNP) (rs1864729) with the lowest P value (P = 3.49E-08), mapped to chromosome 8 near TSPYL5. We also identified 17 imputed SNPs in or near TSPYL5 with P values < 5E-08, one of which, rs2583506, created a functional estrogen response element. We then used a panel of lymphoblastoid cell lines (LCLs) stably transfected with ERα with known genome-wide SNP genotypes to demonstrate that TSPYL5 expression increased after E2 exposure of cells heterozygous for variant TSPYL5 SNP genotypes, but not in those homozygous for wild-type alleles. TSPYL5 knockdown decreased, and overexpression increased aromatase (CYP19A1) expression in MCF-7 cells, LCLs, and adipocytes through the skin/adipose (I.4) promoter. Chromatin immunoprecipitation assay showed that TSPYL5 bound to the CYP19A1 I.4 promoter. A putative TSPYL5 binding motif was identified in 43 genes, and TSPYL5 appeared to function as a transcription factor for most of those genes. In summary, genome-wide significant SNPs in TSPYL5 were associated with elevated plasma E2 in postmenopausal breast cancer patients. SNP rs2583506 created a functional estrogen response element, and LCLs with variant SNP genotypes displayed increased E2-dependent TSPYL5 expression. TSPYL5 induced CYP19A1 expression and that of many other genes. These studies have revealed a novel mechanism for regulating aromatase expression and plasma E2 concentrations in postmenopausal women with ER(+) breast cancer.