Merck
  • Thyrotropin stimulates invasion and growth of follicular thyroid cancer cells via PKC- rather than PKA-activation.
Thyrotropin stimulates invasion and growth of follicular thyroid cancer cells via PKC- rather than PKA-activation.
Biochemical and biophysical research communications (1993-09-30)
T Hoelting, S Tezelman, A E Siperstein, Q Y Duh, O H Clark
ABSTRACT
The signal transduction of TSH in invasion and growth of FTC 133, a human follicular thyroid cancer cell line, was investigated. TSH (0.01-1 mIU/ml) stimulated invasion of FTC 133 by 21% and growth by 20% of basal. Cyclic AMP-stimulators and inhibitors had no effect at any concentration. The PKC-agonist TPA enhanced invasion and growth by 15%, whereas staurosporine, a PKC-antagonist, inhibited them by 32% and 60%, respectively. The latter also reversed TSH stimulation. EGF enhanced invasion (42%) and growth of FTC 133 (25%). Staurosporine did not reverse EGF stimulation. The tyrosine kinase antagonist genistein reversed EGF, but not TSH stimulation. Pertussis toxin inhibited invasion (18%) and growth (22%). Cholera toxin was less inhibitive. We demonstrated for the first time, that TSH stimulates invasion and growth of human thyroid cancer cells in vitro by PKC- rather than PKA-stimulation.