Merck
Characterization of a novel rabbit model of Peyronie's disease.
International journal of impotence research (2023-02-14)
Gokhan Gundogdu, Travis Nguyen, Aarthi Namasivayam, Stephanie Starek, Joel Gelman, Joshua R Mauney
ABSTRACT
Peyronie's disease (PD) is a debilitating pathology which is associated with penile curvature and erectile dysfunction due to the formation of fibrotic plaques in the penile tunica albuginea. In the present study, we developed a novel rabbit model of PD via subtunical injection of recombinant transforming growth factor (TGF)-β1 protein and characterized erectile function and histopathological endpoints following plaque formation. Ten adult male, New Zealand white rabbits were randomized into 3 experimental groups including nonsurgical controls (NSC, N = 3) and those receiving subtunical injections of vehicle (N = 3) or TGF-β1 protein (0.5 µg/50 µl; N = 4). Following 1 month post-op, focal fibrous plaques composed of disorganized collagen type I and III bundles as well as fragmented elastin fibers at TGF-β1 injection sites were observed in contrast to control groups. Cavernosometric and cavernosographic evaluations revealed no significant differences in maximum intracorporal pressures or substantial curvature during papaverine-induced erection in either the vehicle or TGF-β1 cohorts. Immunohistochemical and histomorphometric analyses demonstrated significant increases in elastase 2B expression in TGF-β1-induced plaques as well as significant declines in matrix metalloproteinase (MMP)-2 and -9 expression relative to control levels. Our results demonstrate that PD-like fibrotic plaques can be created in the rabbit penile tunica albuginea following TGF-β1 injection.