Merck
  • Neoplastic ICAM-1 protects lung carcinoma from apoptosis through ligation of fibrinogen.
Neoplastic ICAM-1 protects lung carcinoma from apoptosis through ligation of fibrinogen.
Cell death & disease (2024-08-22)
ShiHui Wang, JunLei Wang, Cui Liu, Lei Yang, XuanQian Tan, ShiYang Chen, Yun Xue, HongBin Ji, GaoXiang Ge, JianFeng Chen
ABSTRACT
Intercellular cell adhesion molecule-1 (ICAM-1) is frequently overexpressed in non-small cell lung cancer (NSCLC) and associated with poor prognosis. However, the mechanism underlying the negative effects of neoplastic ICAM-1 remains obscure. Herein, we demonstrate that the survival of NSCLC cells but not normal human bronchial epithelial cells requires an anti-apoptosis signal triggered by fibrinogen γ chain (FGG)-ICAM-1 interaction. ICAM-1-FGG ligation preserves the tyrosine phosphorylation of ICAM-1 cytoplasmic domain and its association with SHP-2, and subsequently promotes Akt and ERK1/2 activation but suppresses JNK and p38 activation. Abolishing ICAM-1-FGG interaction induces NSCLC cell death by activating caspase-9/3 and significantly inhibits tumor development in a mouse xenograft model. Finally, we developed a monoclonal antibody against ICAM-1-FGG binding motif, which blocks ICAM-1‒FGG interaction and effectively suppresses NSCLC cell survival in vitro and tumor growth in vivo. Thus, suppressing ICAM-1-FGG axis provides a potential strategy for NSCLC targeted therapy.