Merck
  • Fasting-induced activity changes in MC3R neurons of the paraventricular nucleus of the thalamus.
Fasting-induced activity changes in MC3R neurons of the paraventricular nucleus of the thalamus.
Life science alliance (2024-08-07)
Robert A Chesters, Jiajie Zhu, Bethany M Coull, David Baidoe-Ansah, Lea Baumer, Lydia Palm, Niklas Klinghammer, Seve Chen, Anneke Hahm, Selma Yagoub, Lídia Cantacorps, Daniel Bernardi, Katrin Ritter, Rachel N Lippert
ABSTRACT
The brain controls energy homeostasis by regulating food intake through signaling within the melanocortin system. Whilst we understand the role of the hypothalamus within this system, how extra-hypothalamic brain regions are involved in controlling energy balance remains unclear. Here we show that the melanocortin 3 receptor (MC3R) is expressed in the paraventricular nucleus of the thalamus (PVT). We tested whether fasting would change the activity of MC3R neurons in this region by assessing the levels of c-Fos and pCREB as neuronal activity markers. We determined that overnight fasting causes a significant reduction in pCREB levels within PVT-MC3R neurons. We then questioned whether perturbation of MC3R signaling, during fasting, would result in altered refeeding. Using chemogenetic approaches, we show that modulation of MC3R activity, during the fasting period, does not impact body weight regain or total food intake in the refeeding period. However, we did observe significant differences in the pattern of feeding-related behavior. These findings suggest that the PVT is a region where MC3R neurons respond to energy deprivation and modulate refeeding behavior.