Merck
  • Circ_0004140 promotes lung adenocarcinoma progression by upregulating NOVA2 via sponging miR-330-5p.
Circ_0004140 promotes lung adenocarcinoma progression by upregulating NOVA2 via sponging miR-330-5p.
Thoracic cancer (2023-11-03)
Fan Xia, Mei Xie, Jinqi He, Deyun Cheng
ABSTRACT
Circular RNAs (circRNAs) play a significant role in the tumorigenesis and progression of diverse human cancers, including lung adenocarcinoma. A previous study suggested that circ_0004140 expression was increased in lung adenocarcinoma cells. However, the molecular mechanism of circRNA circ_0004140 involved in lung adenocarcinoma is poorly defined. Circ_0004140, microRNA-330-5p (miR-330-5p), and NOVA alternative splicing regulator 2 (NOVA2) expression were determined by real-time quantitative polymerase chain reaction (RT-qPCR). Cell proliferation, apoptosis, migration, invasion, and angiogenesis ability were assessed using 5-ethynyl-2'-deoxyuridine (EdU), flow cytometry, wound healing, transwell, and capillary-like network formation assays. Proliferating cell nuclear antigen (PCNA), cyclin D1, and NOVA2 protein levels were detected using Western blot assay. The interaction between miR-330-5p and circ_0004140 or NOVA2 was verified by dual-luciferase reporter assay. Xenograft tumor model was utilized to assess the role of circ_0004140 in tumor growth in vivo. Circ_0004140 was upregulated in lung adenocarcinoma tissues and cell lines. Circ_0004140 silencing suppressed cell proliferation, migration, invasion and tube formation ability, and triggered the apoptosis of lung adenocarcinoma cells. Circ_0004140 acted as a molecular sponge for miR-330-5p, and miR-330-5p silencing largely reversed circ_0004140 knockdown-induced effects in lung adenocarcinoma cells. NOVA2 was a target of miR-330-5p, and NOVA2 overexpression might largely overturn miR-330-5p overexpression-induced influences in lung adenocarcinoma cells. Circ_0004140 upregulated NOVA2 expression via sponging miR-330-5p in lung adenocarcinoma cells. Circ_0004140 silencing restrained xenograft tumor growth in vivo. Circ_0004140 knockdown might suppress the malignant biological behaviors of lung adenocarcinoma cells via miR-330-5p-dependent regulation of NOVA2.