Merck
  • The immunomodulatory role of paracrine signalling factor VSIG4 in peritoneal metastases.
The immunomodulatory role of paracrine signalling factor VSIG4 in peritoneal metastases.
Scientific reports (2024-07-31)
Yik Yan Chong, Sasinthiran Thiagarajan, Qiu Xuan Tan, Hui Jun Lim, Joey Wee-Shan Tan, Josephine Hendrikson, Gillian Ng, Ying Liu, Clara Yieh Lin Chong, Wanyu Guo, Nye Thane Ngo, Wei-Qiang Leow, Tracy Loh, Xin Xiu Sam, Tony Kiat Hon Lim, Mingzhe Cai, Chin Jin Seo, Jolene Si Min Wong, Khee Chee Soo, Claramae Shulyn Chia, Nicholas Brian Shannon, Chin-Ann Johnny Ong
ABSTRACT
Peritoneal metastasis (PM), the regional progression of intra-abdominal malignancies, is a common sequelae of colorectal cancer (CRC). Immunotherapy is slated to be effective in generating long-lasting anti-tumour response as it utilizes the specificity and memory of the immune system. In the tumour microenvironment, tumour associated macrophages (TAMs) are posited to create an anti-inflammatory pro-tumorigenic environment. In this paper, we aimed to identify immunomodulatory factors associated with colorectal PM (CPM). A publicly available colorectal single cell database (GSE183916) was analysed to identify possible immunological markers that are associated with the activation of macrophages in cancers. Immunohistochemical analysis for V-set and immunoglobin containing domain 4 (VSIG4) expression was performed on tumour microarrays (TMAs) of tumours of colorectal origin (n = 211). Expression of VSIG4 in cell-free ascites obtained from CPM patients (n = 39) was determined using enzyme-linked immunosorbent assay (ELISA). CD163-positive TAMs cluster expression was extracted from a publicly available single cell database and evaluated for the top 100 genes. From these macrophage-expressed genes, VSIG4, a membrane protein produced by the M2 macrophages, mediates the up-regulation of anti-inflammatory and down-regulation of pro-inflammatory macrophages, contributing to an overall anti-inflammatory state. CRC TMA IHC staining showed that low expression of VSIG4 in stromal tissues of primary CRC are associated with poor prognosis (p = 0.0226). CPM ascites also contained varying concentrations of VSIG4, which points to a possible role of VSIG4 in the ascites. The contribution of VSIG4 to CPM development can be further evaluated for its potential as an immunotherapeutic agent.