Merck
  • Single-cell RNA-seq of in vitro expanded cells from cranial neural crest reveals a rare odontogenic sub-population.
Single-cell RNA-seq of in vitro expanded cells from cranial neural crest reveals a rare odontogenic sub-population.
Cell proliferation (2024-01-10)
Yifan Zhao, Shubin Chen, Xiaobo Liu, Xiaoming Chen, Dandan Yang, Jiashu Zhang, Di Wu, Yanmei Zhang, Si Xie, Xiaomei Li, Zhiyuan Wang, Bo Feng, Dajiang Qin, Duanqing Pei, Yaofeng Wang, Jinglei Cai
ABSTRACT
Ecto-mesenchymal cells of mammalian tooth germ develops from cranial neural crest cells. These cells are recognised as a promising source for tooth development and regeneration. Despite the high heterogeneity of the neural crest, the cellular landscape of in vitro cultured cranial neural crest cells (CNCCs) for odontogenesis remains unclear. In this study, we used large-scale single-cell RNA sequencing to analyse the cellular landscape of in vitro cultured mouse CNCCs for odontogenesis. We revealed distinct cell trajectories from primary cells to passage 5 and identified a rare Alx3+/Barx1+ sub-population in primary CNCCs that differentiated into two odontogenic clusters characterised by the up-regulation of Pax9/Bmp3 and Lhx6/Dmp1. We successfully induced whole tooth-like structures containing enamel, dentin, and pulp under the mouse renal capsule using in vitro cultured cells from both cranial and trunk neural crests with induction rates of 26.7% and 22.1%, respectively. Importantly, we confirmed only cells sorted from odontogenic path can induce tooth-like structures. Cell cycle and DNA replication genes were concomitantly upregulated in the cultured NCCs of the tooth induction groups. Our data provide valuable insights into the cell heterogeneity of in vitro cultured CNCCs and their potential as a source for tooth regeneration.