Merck
  • Targeting the Tumor Microenvironment through mTOR Inhibition and Chemotherapy as Induction Therapy for Locally Advanced Head and Neck Squamous Cell Carcinoma: The CAPRA Study.
Targeting the Tumor Microenvironment through mTOR Inhibition and Chemotherapy as Induction Therapy for Locally Advanced Head and Neck Squamous Cell Carcinoma: The CAPRA Study.
Cancers (2022-09-24)
Diane Evrard, Clément Dumont, Michel Gatineau, Jean-Pierre Delord, Jérôme Fayette, Chantal Dreyer, Annemilaï Tijeras-Raballand, Armand de Gramont, Jean-François Delattre, Muriel Granier, Nasredine Aissat, Marie-Line Garcia-Larnicol, Khemaies Slimane, Benoist Chibaudel, Eric Raymond, Christophe Le Tourneau, Sandrine Faivre
ABSTRACT
Mammalian target of rapamycin (mTOR) regulates cellular functions by integrating intracellular signals and signals from the tumor microenvironment (TME). The PI3K-AKT-mTOR pathway is activated in 70% of head and neck squamous cell carcinoma (HNSCC) and associated with poor prognosis. This phase I-II study investigated the effect of mTOR inhibition using weekly everolimus (30 mg for dose level 1, 50 mg for dose level 2) combined with weekly induction chemotherapy (AUC2 carboplatin and 60 mg/m2 paclitaxel) in treatment-naïve patients with locally advanced T3-4/N0-3 HNSCC. Patients received 9 weekly cycles before chemoradiotherapy. Objectives were safety and antitumor activity along with tissue and blood molecular biomarkers. A total of 50 patients were enrolled. Among 41 evaluable patients treated at the recommended dose of 50 mg everolimus weekly, tolerance was good and overall response rate was 75.6%, including 20 major responses (≥50% reduction in tumor size). A significant decrease in expression of p-S6K (p-value: 0.007) and Ki67 (p-value: 0.01) was observed in post-treatment tumor tissue. Pro-immunogenic cytokine release (Th1 cytokines IFN-γ, IL-2, and TNF-β) was observed in the peripheral blood. The combination of everolimus and chemotherapy in HNSCC was safe and achieved major tumor responses. This strategy favorably impacts the TME and might be combined with immunotherapeutic agents.