Merck
  • New Functional Motifs for the Targeted Localization of Proteins to the Nucleolus in Drosophila and Human Cells.
New Functional Motifs for the Targeted Localization of Proteins to the Nucleolus in Drosophila and Human Cells.
International journal of molecular sciences (2024-01-27)
Anna A Ogienko, Mariya O Korepina, Alexey V Pindyurin, Evgeniya S Omelina
ABSTRACT
The nucleolus is a significant nuclear organelle that is primarily known for its role in ribosome biogenesis. However, emerging evidence suggests that the nucleolus may have additional functions. Particularly, it is involved in the organization of the three-dimensional structure of the genome. The nucleolus acts as a platform for the clustering of repressed chromatin, although this process is not yet fully understood, especially in the context of Drosophila. One way to study the regions of the genome that cluster near the nucleolus in Drosophila demands the identification of a reliable nucleolus-localizing signal (NoLS) motif(s) that can highly specifically recruit the protein of interest to the nucleolus. Here, we tested a series of various NoLS motifs from proteins of different species, as well as some of their combinations, for the ability to drive the nucleolar localization of the chimeric H2B-GFP protein. Several short motifs were found to effectively localize the H2B-GFP protein to the nucleolus in over 40% of transfected Drosophila S2 cells. Furthermore, it was demonstrated that NoLS motifs derived from Drosophila proteins exhibited greater efficiency compared to that of those from other species.