Merck
Y-Box-Binding Proteins Have a Dual Impact on Cellular Translation.
International journal of molecular sciences (2024-02-10)
Irina A Eliseeva, Andrey I Buyan, Egor A Smolin, Karina S Kaliadzenka, Sergey Popov, Ivan V Kulakovskiy, Dmitry N Lyabin
ABSTRACT
Y-box-binding proteins (YB proteins) are multifunctional DNA- and RNA-binding proteins that play an important role in the regulation of gene expression. The high homology of their cold shock domains and the similarity between their long, unstructured C-terminal domains suggest that Y-box-binding proteins may have similar functions in a cell. Here, we consider the functional interchangeability of the somatic YB proteins YB-1 and YB-3. RNA-seq and Ribo-seq are used to track changes in the mRNA abundance or mRNA translation in HEK293T cells solely expressing YB-1, YB-3, or neither of them. We show that YB proteins have a dual effect on translation. Although the expression of YB proteins stimulates global translation, YB-1 and YB-3 inhibit the translation of their direct CLIP-identified mRNA targets. The impact of YB-1 and YB-3 on the translation of their mRNA targets is similar, which suggests that they can substitute each other in inhibiting the translation of their mRNA targets in HEK293T cells.