Merck
  • Collagen type IV alpha 1 chain (COL4A1) expression in the developing human lung.
Collagen type IV alpha 1 chain (COL4A1) expression in the developing human lung.
BMC pulmonary medicine (2024-02-09)
Laszlo Markasz, Hamid Mobini-Far, Richard Sindelar
ABSTRACT
Collagen type IV alpha 1 chain (COL4A1) in the basement membrane is an important component during lung development, as suggested from animal models where COL4A1 has been shown to regulate alveolarization and angiogenesis. Less is known about its role in human lung development. Our aim was to study COL4A1 expression in preterm infants with different lung maturational and clinical features. COL4A1 expression in 115 lung samples from newborn infants (21-41 weeks' gestational age; 0-228 days' postnatal age [PNA]) was studied by immunohistochemistry combined with digital image analysis. Cluster analysis was performed to find subgroups according to immunohistologic and clinical data. Patients were automatically categorized into 4 Groups depending on their COL4A1 expression. Expression of COL4A1 was mainly extracellular in Group 1, low in Group 2, intracellular in Group 3, and both extra- and intracellular in Group 4. Intracellular/extracellular ratio of COL4A1 expression related to PNA showed a distinctive postnatal maturational pattern on days 1-7, where intracellular expression of COL4A1 was overrepresented in extremely preterm infants. COL4A1 expression seems to be highly dynamic during the postnatal life due to a possible rapid remodeling of the basement membrane. Intracellular accumulation of COL4A1 in the lungs of extremely premature infants occurs more frequently between 1 and 7 postnatal days than during the first 24 hours. In view of the lung arrest described in extremely preterm infants, the pathological and/or developmental role of postnatally increased intracellular COL4A1 as marker for basement membrane turnover, needs to be further investigated.