Merck
Human transferrin receptor can mediate SARS-CoV-2 infection.
Proceedings of the National Academy of Sciences of the United States of America (2024-02-26)
Zhiyi Liao, Chaoming Wang, Xiaopeng Tang, Mengli Yang, Zilei Duan, Lei Liu, Shuaiyao Lu, Lei Ma, Ruomei Cheng, Gan Wang, Hongqi Liu, Shuo Yang, Jingwen Xu, Dawit Adisu Tadese, James Mwangi, Peter Muiruri Kamau, Zhiye Zhang, Lian Yang, Guoyang Liao, Xudong Zhao, Xiaozhong Peng, Ren Lai
ABSTRACT
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection has been detected in almost all organs of coronavirus disease-19 patients, although some organs do not express angiotensin-converting enzyme-2 (ACE2), a known receptor of SARS-CoV-2, implying the presence of alternative receptors and/or co-receptors. Here, we show that the ubiquitously distributed human transferrin receptor (TfR), which binds to diferric transferrin to traffic between membrane and endosome for the iron delivery cycle, can ACE2-independently mediate SARS-CoV-2 infection. Human, not mouse TfR, interacts with Spike protein with a high affinity (KD ~2.95 nM) to mediate SARS-CoV-2 endocytosis. TfR knock-down (TfR-deficiency is lethal) and overexpression inhibit and promote SARS-CoV-2 infection, respectively. Humanized TfR expression enables SARS-CoV-2 infection in baby hamster kidney cells and C57 mice, which are known to be insusceptible to the virus infection. Soluble TfR, Tf, designed peptides blocking TfR-Spike interaction and anti-TfR antibody show significant anti-COVID-19 effects in cell and monkey models. Collectively, this report indicates that TfR is a receptor/co-receptor of SARS-CoV-2 mediating SARS-CoV-2 entry and infectivity by likely using the TfR trafficking pathway.