Merck
  • Integrating iron metabolism-related gene signature to evaluate prognosis and immune infiltration in nasopharyngeal carcinoma.
Integrating iron metabolism-related gene signature to evaluate prognosis and immune infiltration in nasopharyngeal carcinoma.
Discover oncology (2024-04-11)
Jiaming Su, Guanlin Zhong, Weiling Qin, Lu Zhou, Jiemei Ye, Yinxing Ye, Chang Chen, Pan Liang, Weilin Zhao, Xue Xiao, Wensheng Wen, Wenqi Luo, Xiaoying Zhou, Zhe Zhang, Yonglin Cai, Cheng Li
ABSTRACT
Dysregulation of iron metabolism has been shown to have significant implications for cancer development. We aimed to investigate the prognostic and immunological significance of iron metabolism-related genes (IMRGs) in nasopharyngeal carcinoma (NPC). Multiple Gene Expression Omnibus (GEO) and The Cancer Genome Atlas (TCGA) datasets were analyzed to identify key IMRGs associated with prognosis. Additionally, the immunological significance of IMRGs was explored. A novel risk model was established using the LASSO regression algorithm, incorporating three genes (TFRC, SLC39A14, and ATP6V0D1).This model categorized patients into low and high-risk groups, and Kaplan-Meier analysis revealed significantly shorter progression-free survival for the high-risk group (P < 0.0001). The prognostic model's accuracy was additionally confirmed by employing time-dependent Receiver Operating Characteristic (ROC) curves and conducting Decision Curve Analysis (DCA). High-risk patients were found to correlate with advanced clinical stages, specific tumor microenvironment subtypes, and distinct morphologies. ESTIMATE analysis demonstrated a significant inverse relationship between increased immune, stromal, and ESTIMATE scores and lowered risk score. Immune analysis indicated a negative correlation between high-risk score and the abundance of most tumor-infiltrating immune cells, including dendritic cells, CD8+ T cells, CD4+ T cells, and B cells. This correlation extended to immune checkpoint genes such as PDCD1, CTLA4, TIGIT, LAG3, and BTLA. The protein expression patterns of selected genes in clinical NPC samples were validated through immunohistochemistry. This study presents a prognostic model utilizing IMRGs in NPC, which could assist in assessing patient prognosis and provide insights into new therapeutic targets for NPC.